Skip to Content
Merck
All Photos(1)

Key Documents

E-038

Supelco

Ethanol-100 (5 ampules/kit)

100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®

Synonym(s):

Ethanol solution, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 5 × 5 mL

manufacturer/tradename

Cerilliant®

concentration

100 mg/dL in H2O

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A kit of NIST traceable ethanol standards suitable for calibration of ethanol testing methods. These Certified Reference Materials are widely used in forensic analysis and clinical toxicology applications for determination of blood alcohol content.

Application

The certified reference solution (CRM) in solution can also be used as follows:

  • Analysis of ethanol content by proton nuclear magnetic resonance (1H NMR) spectroscopy in whole blood samples[1]
  • Development of a paper-based biosensor for directly measuring ethanol content in whole blood based on enzymatic colorimetric assessment combined with in-situ headspace gas separation
  • Detection and quantitative analysis of ethanol and other volatile organic compounds (VOCs) in bovine blood samples by gas chromatography coupled to vacuum ultraviolet spectroscopy (GC-VUV)
  • Development and validation of an analytical method based on head space-gas chromatography (HS-GC) combined with flame ionization detector (FID) to quantify ethanol in blood samples[2]

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of ethanol in blood using headspace gas chromatography with flameionization detector (HS-GC-FID): Validation of a method
Mihretu LD, et al.
Cogent Chemistry , 6, 1760187-1760187 (2020)
Alternative determination of blood alcohol concentration by 1H NMR spectroscopy
Zailer E and Diehl BWK
Journal of Pharmaceutical and Biomedical Analysis, 119, 59-64 (2016)
S Enocksson et al.
The Journal of clinical investigation, 95(5), 2239-2245 (1995-05-01)
Although it is well established in several mammalian species that beta 3-adrenoceptors play a major role in regulating lipolysis and thermogenesis in adipose tissue, the functional existence and role of this receptor subtype in man has been controversial. We investigated
Thiago M Pais et al.
PLoS genetics, 9(6), e1003548-e1003548 (2013-06-12)
The yeast Saccharomyces cerevisiae is able to accumulate ≥17% ethanol (v/v) by fermentation in the absence of cell proliferation. The genetic basis of this unique capacity is unknown. Up to now, all research has focused on tolerance of yeast cell
A Nyéki et al.
Journal of chromatography. B, Biomedical sciences and applications, 755(1-2), 73-84 (2001-06-08)
Urinary metabolic ratios of caffeine are used in humans to assess the enzymatic activities of cytochrome P450 isoenzyme 1A2 (CYP1A2), xanthine oxidase (XO) and for phenotyping individuals for the bimodal N-acetyltransferase 2 (NAT2), all of them involved in the activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service