Skip to Content
Merck
All Photos(1)

Documents

E-064

Supelco

Ethyl sulfate sodium salt

1.0 mg/mL in methanol (as ethyl sulfate), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5O4SNa
CAS Number:
Molecular Weight:
148.11
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as ethyl sulfate)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[Na+].CCOS([O-])(=O)=O

InChI

1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1

InChI key

RRLOOYQHUHGIRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine alchohol testing. Ethyl sulfate is a primary urinary metabolite of consumed alcohol and is indicative of recent alcohol consumption.

Application


  • Ethyl sulfate sodium salt in pharmacological studies: This compound is instrumental in exploring the pharmacological properties of Musa paradisiaca, demonstrating its utility in the assessment of anti-inflammatory, analgesic, and wound healing properties in preclinical studies. This research highlights the adaptability of ethyl sulfate sodium salt in complex biological systems, offering broad implications for therapeutic applications (Ekweogu et al., 2024).

  • Stable isotope-labeled ethyl sulfate sodium salt: Leveraging its stable isotopic properties, ethyl sulfate sodium salt is critical in membrane research, particularly in studies focused on the microscopic observation of membrane fusion mechanisms. Its use facilitates precise biochemical interactions and understanding of viral entry processes, enhancing virological studies and potential therapeutic targeting (Nishigami et al., 2023).

  • Ethyl sulfate sodium salt in environmental research: Utilized in a study to enhance the resistance of microorganisms to heavy metals, ethyl sulfate sodium salt proves effective in bioremediation research. Its application assists in the development of more resilient biological agents for environmental cleanup, showcasing its crucial role in ecological sciences (Liu et al., 2017).

  • Research use in energy efficiency: Ethyl sulfate sodium salt is employed in innovative studies aiming to develop more energy-efficient processes for biomass processing. Its role in mediator behavior in ionic liquids exemplifies its contribution to advancing green chemistry and sustainable industrial practices (Eshtaya et al., 2016).

  • Ethyl sulfate sodium salt in atmospheric science: This compound is crucial in studies exploring the hygroscopic growth of aerosols, impacting our understanding of atmospheric chemistry and climate modeling. Its use in aerosol research underscores its significance in environmental monitoring and climate change studies (Estillore et al., 2016).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service