Key Documents
Z742263
DURAN® GL 56 Bottle Tag
orange silicone
Synonym(s):
292435626
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
orange silicone
packaging
pack of 20 ea
manufacturer/tradename
DURAN® 292435626
Related Categories
General description
Bottle tags can be used on their own for color identification or to secure the protective light sheild around the bottle. The GL 56 bottle tags are available in four colors (orange, yellow, blue and purple).
Features and Benefits
  • Manufactured from silicone rubber
  • The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
  • The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
  • Feature a graspable tab that makes them easy to remove with bare or gloved hands
  • Chemically robust, heat resistant and reusable
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)