Skip to Content
Merck
All Photos(1)

Key Documents

Z742389

Duran® double walled jacketed GLS 80 bottle

capacity 1000 mL, polypropylene pour ring, blue polypropylene cap

Synonym(s):

242565456, Double Walled GLS 80 Bottle, Duran Jacketed Bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

PTFE connector
blue polypropylene cap
borosilicate glass 3.3 bottle
colorless bottle
polypropylene pour ring
round bottle

Joint

Joints threaded neck (65)

feature

with cap

packaging

pack of 1 ea

manufacturer/tradename

DURAN® 242565456

parameter

-40-120 °C temp. range

H × diam.

270 mm × 110 mm

capacity

1000 mL

connector size

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

The DURAN double walled, wide mouth bottles GLS 80 incorporate an integral jacket that isolates the contents from the external environment. Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN bottle. The DURAN double walled bottles offer a sealable and more flexible alternative to open topped jacketed beakers.

Application

  • Simple, low cost, jacketed chemostat or bioreactor for cell culture.
  • Vessel for temperature-controlled reactions such as enzyme digestions.
  • Simple continuous flow stirred reactor system with the optional GLS 80 cap components.
  • Thermostatic vessel to maintain fluids, or high viscosity liquids at a constant temperature, useful when working with endo- or exothermic reagents.
  • Large cold trap or condenser when used with dry ice in the inner container.

Features and Benefits

  • All components are fully autoclavable
  • The wide 65 mm inner diameter of the GLS 80 wide mouth provides easy access to the contents, or for cleaning.
  • The flat inner base is compatible with magnetic stir bars, or the DURAN GLS 80 magnetic stirrer cap system.
  • Compatible with full range of wide mouth GLS 80 connection cap systems for liquid addition or removal during processing.
  • The inner vessel features an easy-to-read graduated volume scale.
  • The GL 18 outlet hose connectors are removable for increased safety
  • The positions of the 10 mm PTFE inlet/outlet hose connectors are optimized for a uniform flow around the jacket to ensure consistent heating or cooling.

Other Notes

The maximum pressure in the double walled glass jacket must not exceed +50 kPa gauge. These products must not be used under vacuum.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service