Skip to Content
Merck
All Photos(1)

Key Documents

Z530662

Sigma-Aldrich

Aldrich® vacuum manifold with joints

positions, 3, joint: ST/NS 14/20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161501
NACRES:
NB.79

Pricing and availability is not currently available.

Joint

joint: ST/NS14/20

feature

positions 3

L

300 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
490679490636279455
isotopic purity

≥99 atom % 13C

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C

assay

≥97% (CP)

assay

-

assay

-

assay

-

mass shift

M+3

mass shift

M+1

mass shift

M+3

mass shift

M+1

form

solid

form

solid

form

solid

form

solid

General description

This practical design provides clearance to accommodate 250mL flasks with a snap of a Keck® clip. Manifolds are available with either <ST/>14/20, <ST/>24/40, or <ST/>29/32 joints. Glass stopcocks have a 4mm bore.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Keck is a registered trademark of Adcuram Lab Technologies GmbH

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service