Z414158
Aldrich® dual bank manifold with high-vacuum PTFE valves
standard manifold, positions, 4
All Photos(1)
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
Recommended Products
feature
positions 4
L
400 mm
Related Categories
General description
NOTE: J Young valves are currently unavailable. We are temporarily supplying another high quality valve that Sigma-Aldrich has tested and approved.
The dual bank manifolds come with 0-10 mm bore high-vacuum PTFE valves.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)