Skip to Content
Merck
All Photos(1)

Documents

W373605

Sigma-Aldrich

L-Tyrosine

FG

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
FEMA Number:
3736
Beilstein:
392441
EC Number:
MDL number:
UNSPSC Code:
12164502
eCl@ss:
32160406
PubChem Substance ID:
Flavis number:
17.022
NACRES:
NA.21

biological source

corn

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 173.65

optical activity

[α]20/D −10.6°, c = 4 in 1 M HCl

mp

>300 °C (dec.) (lit.)

solubility

water: soluble 0.479 g/L at 25 °C

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

odorless

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Tyrosine is an important amino acid mainly used as a dietary supplement.
L-tyrosine, an aromatic amino acid, is used common dietary supplement that has a wide range of applications in food and pharmaceutical industry. It is a precursor for preparing industrially important compounds such as 3,4-dihydroxy-L-phenylalanine (L-DOPA), melanin and p-hydroxystyrene. L-tyrosine can be synthesized by different biotechnological methods which are ecofriendly.

Other Notes

Amino acid precursor of dopamine and other catecholamines

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Rahm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 546-550 (2014-02-26)
Low-grade gliomas (LGGs) may harbor malignant foci, which are characterized by increased tumor cellularity and angiogenesis. We used diffusion-weighted MR imaging (apparent diffusion coefficient [ADC]) and PET with the amino acid O-(2-(18)F-fluorethyl)-L-tyrosine ((18)F-FET) to search for focal changes of diffusion
Vincent Dunet et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(2), 207-214 (2012-02-04)
For the past decade, PET with (18)F-fluoro-ethyl-tyrosine ((18)F-FET) has been used in the evaluation of patients with primary brain tumors (PBTs), but so far series have reported only a limited number of patients. The purpose of this systematic review and
Hideki Hara et al.
Nature immunology, 14(12), 1247-1255 (2013-11-05)
The inflammasome adaptor ASC contributes to innate immunity through the activation of caspase-1. Here we found that signaling pathways dependent on the kinases Syk and Jnk were required for the activation of caspase-1 via the ASC-dependent inflammasomes NLRP3 and AIM2.
Keisuke Saito et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7690-7695 (2013-04-20)
Using quantum mechanics/molecular mechanics calculations and the 1.9-Å crystal structure of Photosystem II [Umena Y, Kawakami K, Shen J-R, Kamiya N (2011) Nature 473(7345):55-60], we investigated the H-bonding environment of the redox-active tyrosine D (TyrD) and obtained insights that help
Tina Lütke-Eversloh et al.
Applied microbiology and biotechnology, 77(4), 751-762 (2007-10-31)
The aromatic amino acid L-tyrosine is used as a dietary supplement and has promise as a valuable precursor compound for various industrial and pharmaceutical applications. In contrast to chemical production, biotechnological methods can produce L-tyrosine from biomass feedstocks under environmentally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service