W360902
2-Acetyl-5-methylfuran
≥98%, FG
Synonym(s):
NSC 80404
About This Item
Halal
Kosher
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Kosher
reg. compliance
EU Regulation 1334/2008 & 872/2012
vapor density
>1 (vs air)
Assay
≥98%
refractive index
n20/D 1.512 (lit.)
bp
100-101 °C/25 mmHg (lit.)
density
1.066 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
musty; nutty; sweet
SMILES string
CC(=O)c1ccc(C)o1
InChI
1S/C7H8O2/c1-5-3-4-7(9-5)6(2)8/h3-4H,1-2H3
InChI key
KEFJLCGVTHRGAH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Biochar-extracted liquor stimulates nitrogen related gene expression on improving nitrogen utilization in rice seedling.: This study investigates the use of 2-Acetyl-5-methylfuran derived from biochar-extracted liquor to enhance nitrogen uptake in rice seedlings, showing potential to improve agricultural productivity and environmental sustainability (Gao et al., 2023).
- Occurrence of (suspected) genotoxic flavoring substances in Belgian alcohol-free beers.: This study assesses the presence of 2-Acetyl-5-methylfuran in alcohol-free beers, discussing its implications for food safety and consumer health, highlighting the need for stringent quality control in the beverage industry (Dusart et al., 2022).
- Characterization of the physicochemical changes and volatile compound fingerprinting during the chicken sugar-smoking process.: Research on the role of 2-Acetyl-5-methylfuran in the flavor profile of smoked chicken, providing valuable information for the food industry on enhancing sensory attributes of smoked meats (Chang et al., 2021).
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
176.0 °F - closed cup
Flash Point(C)
80 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service