Skip to Content
Merck
All Photos(1)

Key Documents

W346101

Sigma-Aldrich

2-Isopropylphenol

≥98%, FG

Synonym(s):

2-propan-2-ylphenol, O-cumenol, Prodox 131

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3461
Beilstein:
1363322
EC Number:
Council of Europe no.:
11234
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.044
NACRES:
NA.21
Organoleptic:
medicinal
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA

vapor pressure

<0.05 mmHg ( 25 °C)

Assay

≥98%

refractive index

n20/D 1.526 (lit.)

bp

212-213 °C (lit.)

mp

12-16 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

medicinal

SMILES string

CC(C)c1ccccc1O

InChI

1S/C9H12O/c1-7(2)8-5-3-4-6-9(8)10/h3-7,10H,1-2H3

InChI key

CRBJBYGJVIBWIY-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Reichlin et al.
Applied and environmental microbiology, 60(12), 4587-4591 (1994-12-01)
Pseudomonas sp. strain HBP1 Prp grew on 2-isopropylphenol as the sole carbon and energy source with a maximal specific growth rate of 0.14 h-1 and transient accumulation of isobutyric acid. Oxygen uptake experiments with resting cells and enzyme assays with
Martin Barann et al.
Anesthesia and analgesia, 106(3), 846-857 (2008-02-23)
5-Hydroxytryptamine type 3 (5-HT3) receptors are excitatory ligand-gated ion channels which are involved in postoperative nausea and vomiting. They are depressed by the anesthetic propofol, which, in contrast, enhances the activity of inhibitory ligand-gated ion channels such as gamma-aminobutyric acid
Kevin A Harvey et al.
Bioorganic & medicinal chemistry, 18(5), 1866-1874 (2010-02-16)
The present study describes the characterization and evaluation of novel anticancer conjugates, 2,6-diisopropylphenol-docosahexaenoate (PP-DHA), and its analogues including 2,4-diisopropylphenol-docosahexaenoate (DIPP-DHA), 2-isopropylphenol-docosahexaenoate (IPP-DHA), 2-cyclohexanephenol-docosahexaenoate (CHP-DHA) and phenol-docosahexaenoate (P-DHA) on breast cancer cell lines. Representative breast cancer cell lines, based on estrogen
Yongnian Ni et al.
Talanta, 76(3), 513-521 (2008-07-01)
The binding interaction of the pesticide Isoprocarb and its degradation product, sodium 2-isopropylphenate, with bovine serum albumin (BSA) was studied by spectrofluorimetry under simulated physiological conditions. Both Isoprocarb and sodium 2-isopropylphenate quenched the intrinsic fluorescence of BSA. This quenching proceeded
Andrzej L Dawidowicz et al.
Journal of food science and technology, 52(7), 4523-4529 (2015-07-04)
The presented paper shows the influence of temperature increase in the range typically used during antioxidant measurements (15-35 °C) on the estimation of antioxidant properties of phenolic compounds (caffeic acid, ferulic acid, gallic acid, trolox, butylhydroxyanisole, butylhydroxytoluene and 2,6-diisopropylphenol) in associating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service