Skip to Content
Merck
All Photos(2)

Key Documents

W360000

Sigma-Aldrich

4-(Methylthio)butanol

97%, FG

Synonym(s):

4-(Methylthio)-1-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3S(CH2)4OH
CAS Number:
Molecular Weight:
120.21
FEMA Number:
3600
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.078
NACRES:
NA.21
Organoleptic:
cabbage; alliaceous; sulfurous
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

97%

refractive index

n20/D 1.487 (lit.)

bp

95-98 °C/20 mmHg (lit.)

density

0.993 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cabbage; alliaceous; sulfurous

SMILES string

CSCCCCO

InChI

1S/C5H12OS/c1-7-5-3-2-4-6/h6H,2-5H2,1H3

InChI key

JNTVUHZXIJFHAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service