Skip to Content
Merck
All Photos(1)

Documents

W330906

Sigma-Aldrich

2-Methylpyrazine

≥99%, FCC, FG

Synonym(s):

2-methyl-1,4-diazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
FEMA Number:
3309
Beilstein:
105778
EC Number:
Council of Europe no.:
2270
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.027
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥99%

refractive index

n20/D 1.504 (lit.)

bp

135 °C/761 mmHg (lit.)

mp

−29 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

musty; nutty; peanut

SMILES string

Cc1cnccn1

InChI

1S/C5H6N2/c1-5-4-6-2-3-7-5/h2-4H,1H3

InChI key

CAWHJQAVHZEVTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylpyrazine is one of the major flavor volatiles reported in rice cakes, roasted peanuts and soybean products.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoyu (soy sauce) volatile flavor components: basic fraction.
Nunomura N, et al.
Journal of Agricultural and Food Chemistry, 42(11), 2123-2128 (1978)
Volatile flavor components of deep fat-fried soybeans.
Wilkens WF & Lin FM.
Journal of Agricultural and Food Chemistry, 18(3), 337-339 (1970)
Pyrazine contents and oxidative stabilities of roasted soybean oils.
Jung MY, et al.
Food Chemistry, 60(1), 95-102 (1997)
Odor threshold levels of pyrazine compounds and assessment of their role in the flavor of roasted foods.
Koehler PE, et al.
Journal of Food Science, 36(5), 816-818 (1971)
`Flavor?fade?and Off?Flavors in Ground Roasted Peanuts As Related to Selected Pyrazines and Aldehydes
Warner KJH, et al.
Journal of Food Science, 61(2), 469-472 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service