Skip to Content
Merck
All Photos(1)

Key Documents

W318809

Sigma-Aldrich

2-Methyl-3-furanthiol

≥95%, FG

Synonym(s):

2-methyl-3-furyl mercaptan, Fish thiol, Oxy cyclothione-030

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
FEMA Number:
3188
EC Number:
Council of Europe no.:
11678
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.055
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

300
400

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

vapor density

>1 (vs air)

Assay

≥95%

refractive index

n20/D 1.518 (lit.)

bp

57-60 °C/44 mmHg (lit.)

density

1.145 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fishy; meaty; metallic; sulfurous

SMILES string

Cc1occc1S

InChI

1S/C5H6OS/c1-4-5(7)2-3-6-4/h2-3,7H,1H3

InChI key

RUYNUXHHUVUINQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-3-furanthiol, a sulfur-containing compound, is the key odorant in cooked ham, wine and canned tuna fish.

Application


  • Elucidating salt-reduction mechanisms of aroma-active compounds from yeast extracts through sensomics approaches and electroencephalography.: This innovative study leverages both sensory science and neurotechnology to uncover how 2-methyl-3-furanthiol and other key compounds in yeast extracts can enhance flavor profiles while reducing sodium content in foods, aligning with health trends and consumer demands for lower-sodium options (Shan et al., 2024).

  • Widely Targeted Metabolomics and Network Pharmacology Reveal the Nutritional Potential of Yellowhorn (Xanthoceras sorbifolium Bunge) Leaves and Flowers.: This study demonstrates the use of targeted metabolomics to profile bioactive compounds, including 2-methyl-3-furanthiol, in Yellowhorn plant parts, which are emerging as a nutritional powerhouse with potential health benefits (Sha et al., 2024).

  • Metabolomics reveals factors affecting the radical reaction of sulfides during thermal processing for meaty aroma.: This research identifies how 2-methyl-3-furanthiol and other sulfides interact during cooking to create rich, meaty flavors, providing valuable insights for the culinary industry on optimizing flavor profiles in cooked meats (Zhang et al., 2024).

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of 2-Methyl-3-Furanthiol in the Steam Distillate from Canned Tuna Fish.
WITHYCOMBE D and MUSSINAN CYJ.
Journal of Food Science, 53(2), 658-658 (1988)
Mosciano, G.
Perfumer & Flavorist, 26, 58-58 (2001)
A novel method for quantification of 2-methyl-3-furanthiol and 2-furanmethanethiol in wines made from Vitis vinifera grape varieties.
Tominaga T & Dubourdieu D
Journal of Agricultural and Food Chemistry, 54(1), 29-33 (2006)
Caroline Thomas et al.
Food chemistry, 155, 207-213 (2014-03-07)
The aim of this work was to identify and gain further knowledge on the origin of sulfur compounds present in the volatile fraction of cooked ham, and on their role in the aroma of this product. To this end, we
Mosciano, G.
Perfumer & Flavorist, 21, 53-53 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service