Skip to Content
Merck
All Photos(1)

Key Documents

441163

Sigma-Aldrich

2-Methyl-3-furanthiol

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

>1 (vs air)

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

57-60 °C/44 mmHg (lit.)

density

1.145 g/mL at 25 °C

SMILES string

Cc1occc1S

InChI

1S/C5H6OS/c1-4-5(7)2-3-6-4/h2-3,7H,1H3

InChI key

RUYNUXHHUVUINQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of 2-methyl-3-furanthiol, 2-furfurylthiol, 3-mercapto-2-pentanone, and 2-mercapto-3-pentanone in heated meat.
Kerscher R and Grosch W.
Journal of Agricultural and Food Chemistry, 46(5), 1954-1958 (1998)
Caroline Thomas et al.
Food chemistry, 173, 790-795 (2014-12-04)
This study shows that thiamine plays a major role in the formation of three key odorants of cooked ham: 2-methyl-3-furanthiol, 2-methyl-3-methyldithiofuran, and bis(2-methyl-3-furyl)disulphide. Analyses revealed that under identical cooking conditions, the productions of these three aroma compounds increase in a
Identification of 2-Methyl-3-Furanthiol in the Steam Distillate from Canned Tuna Fish.
WITHYCOMBE D and MUSSINAN CYJ.
Journal of Food Science, 53(2), 658-658 (1988)
Caroline Thomas et al.
Food chemistry, 155, 207-213 (2014-03-07)
The aim of this work was to identify and gain further knowledge on the origin of sulfur compounds present in the volatile fraction of cooked ham, and on their role in the aroma of this product. To this end, we
Y Bezman et al.
Journal of agricultural and food chemistry, 49(11), 5425-5432 (2001-11-21)
The occurrence of methional in fresh orange juice, and possible occurrence of beta-damascenone in heated orange juice, has been previously suggested. Here we report on the occurrence of 2-methyl-3-furanthiol in the headspace, collected by solid-phase micro-extraction, of fresh, pasteurized, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service