Skip to Content
Merck
All Photos(1)

Key Documents

W305618

Sigma-Aldrich

Tetrahydrofurfuryl alcohol

≥98%

Synonym(s):

NSC 15434, Tetrahydro-2-furanmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
FEMA Number:
3056
Beilstein:
102723
EC Number:
Council of Europe no.:
2029
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.020
NACRES:
NA.21
Organoleptic:
vegetable
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FDA 21 CFR 117

vapor density

3.52 (vs air)

vapor pressure

2.3 mmHg ( 39 °C)

Assay

≥98%

expl. lim.

9.7 %

refractive index

n20/D 1.452 (lit.)

bp

178 °C (lit.)

mp

−80 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

vegetable

SMILES string

OCC1CCCO1

InChI

1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2

InChI key

BSYVTEYKTMYBMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofurfuryl alcohol is one of the aroma constituents of coffee.

Application


  • Electrochemical oxidation of tetrahydrofurfuryl alcohol on boron-doped diamond anode: Influence of current density and electrolyte solution.: Research into the electrochemical pathways of tetrahydrofurfuryl alcohol offers insights into its potential for energy storage applications, focusing on how varying electrical and solvent conditions affect its oxidation rates (Xiong Y et al., 2023).

  • Polar-Functionalized Polyethylenes Enabled by Palladium-Catalyzed Copolymerization of Ethylene and Butadiene/Bio-Based Alcohol-Derived Monomers.: This paper presents a novel approach to the synthesis of functionalized polymers using tetrahydrofurfuryl alcohol derivatives, which could revolutionize materials science with new, sustainable polymer blends (Jian Z et al., 2023).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contribution of coffee aroma constituents to the mutagenicity of coffee.
Aeschbacher HU, et al.
Food And Chemical Toxicology, 27(4), 227-232 (1989)
[Acute poisoning with tetrahydrofurfuryl alcohol combined with methanol].
B F Murashov et al.
Voenno-meditsinskii zhurnal, (10)(10), 32-34 (1983-10-01)
S M Bellm et al.
The Journal of chemical physics, 136(24), 244301-244301 (2012-07-05)
Cross section data for electron scattering from DNA are important for modelling radiation damage in biological systems. Triply differential cross sections for the electron impact ionization of the highest occupied outer valence orbital of tetrahydrofurfuryl alcohol, which can be considered
T Schräder et al.
Journal of bacteriology, 183(24), 7408-7411 (2001-11-22)
Different aldehyde dehydrogenases (AlDHs) were formed during growth of Ralstonia eutropha Bo on tetrahydrofurfuryl alcohol (THFA). One of these enzymes, AlDH 4, was purified and characterized as a homodimer containing no prosthetic groups, showing a strong substrate inhibition, and having
G Zarnt et al.
Journal of bacteriology, 183(6), 1954-1960 (2001-02-27)
The quinohemoprotein tetrahydrofurfuryl alcohol dehydrogenase (THFA-DH) from Ralstonia eutropha strain Bo was investigated for its catalytic properties. The apparent k(cat)/K(m) and K(i) values for several substrates were determined using ferricyanide as an artificial electron acceptor. The highest catalytic efficiency was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service