Skip to Content
Merck
All Photos(1)

Key Documents

T12653

Sigma-Aldrich

Tetrahydrofurfuryl alcohol

98%

Synonym(s):

Oxolan-2-yl methanol, THFA, NSC 15434, Tetrahydro-2-furanmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
Beilstein:
102723
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.52 (vs air)

Quality Level

vapor pressure

2.3 mmHg ( 39 °C)

Assay

98%

expl. lim.

9.7 %

refractive index

n20/D 1.452 (lit.)

bp

178 °C (lit.)

mp

−80 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

OCC1CCCO1

InChI

1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2

InChI key

BSYVTEYKTMYBMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofurfuryl alcohol is a non-carcinogenic, biodegradable, protic solvent.

Application

  • Tetrahydrofurfuryl alcohol (THFA) is a solvent modifier for sodium borohydride used in enantioselective reduction of aromatic imines or ketones to the corresponding alcohols.
  • It can be used to synthesize tetrahydrofurfuryl acetate, tetrahydrofurfuryl propionate and tetrahydrofurfuryl butyrate, which are used as flavoring agents in food industry.
  • Application of THFA as a solvent for water-based colorants; coupling solvent for agricultural formulations such as pesticides, fungicides and herbicides; plasticizer for paints and varnishes are reported.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fenaroli's handbook of flavor ingredients.
Fenaroli's Handbook of Flavor Ingredients (2016)
Encyclopedia of food & color additives.
Encyclopedia of Food and Color Additives (1996)
CLH report: `Tetrahydrofurfuryl alcohol (THFA)?.
unpublished (2011)
Enantioselective borohydride reduction catalyzed by optically active cobalt complexes.
Yamada T, et al.
Chemistry?A European Journal , 9(18), 4485-4509 (2003)
Encyclopedia of food & color additives.
Encyclopedia of Food and Color Additives (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service