W298944
Rose oil, Bulgarian
natural, FG, product of Essential Oils and Herbs Ltd
About This Item
Fragrance grade
natural
Recommended Products
biological source
Rosa damascena
Quality Level
grade
FG
Fragrance grade
natural
Agency
follows IFRA guidelines
reg. compliance
EU 1334/2008 & 178/2002
EU Regulation 1223/2009
FDA 21 CFR 117
description
Fragrance allergen: Methyl and Furfural
composition
Contains IFRA restricted Benzaldehyde, Damascone (alpha, beta), Furfural. EU 1223/2009 restricted Limonene and linalool, and EU 1223/2009 and IFRA restricted Citral, Citronellol, Geraniol, Eugenol, Farnesol, Isoeugenol, Methyl Eugenol and Benzyl Benzoate
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
eugenol, benzyl benzoate, farnesol, citral (neral+geranial), limonene (sum of D, L and DL), isoeugenol, cis-β-damascone, geraniol, citronellol, eugenol, benzaldehyde
greener alternative category
, Aligned
Organoleptic
honey; floral; waxy; rose
Related Categories
General description
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
150.8 °F - closed cup
Flash Point(C)
66 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service