Skip to Content
Merck
All Photos(2)

Key Documents

I17206

Sigma-Aldrich

Isoeugenol

98%, mixture of cis and trans

Synonym(s):

2-Methoxy-4-propenylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH=CHCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1909602
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

98%

refractive index

n20/D 1.575 (lit.)

bp

132 °C/10 mmHg (lit.)

mp

27 °C (lit.)

density

1.077 g/mL at 25 °C

SMILES string

OC1=CC=C(/C=C/C)C=C1OC

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3-7,11H,1-2H3/b4-3+

InChI key

BJIOGJUNALELMI-ONEGZZNKSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Frahm et al.
Animals : an open access journal from MDPI, 10(7) (2020-07-24)
In this preliminary study, we compared changes in mechanical nociceptive thresholds (MNT) of calves and goat kids injected with clove oil or isoeugenol under the horn bud as a potential, more welfare-friendly alternative to hot-iron disbudding. Twenty male calves and
Valentina Galbiati et al.
Archives of toxicology, 86(2), 239-248 (2011-10-05)
We previously demonstrated in the human promyelocytic cell line THP-1 that all allergens tested, with the exception of the prohapten isoeugenol, induced a dose-related release of interleukin-8 (IL-8). In the present study, we investigated whether this abnormal behavior was regulated
Michaela Kalmes et al.
Journal of toxicology and environmental health. Part A, 75(8-10), 478-491 (2012-06-13)
The phenolic derivatives eugenol and isoeugenol, which are naturally found in essential oils of different spices, are commonly used as fragrances. Recently data demonstrated that growth suppression produced by these substances occurs in keratinocytes and that the effects may be
Batirtze Prats-Mateu et al.
Scientific reports, 8(1), 11804-11804 (2018-08-09)
Raman microscopy is a powerful imaging technique for biological materials providing information about chemistry in context with microstructure. A 532 nm laser is often used as excitation source, because high spatial resolution and signal intensity can be achieved. The latter can
T Atsumi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 19(8), 1025-1033 (2005-06-21)
Eugenol (compound in , 4-allyl-2-methyoxyphenol) and isoeugenol (compound in , 4-propenyl-2-methoxyphenol), both used as a flavor agent in cosmetic and food products, have both prooxidant and antioxidant activities. Their adverse effects such as allergic and inflammatory reaction may be due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service