Skip to Content
Merck
All Photos(1)

Key Documents

W273309

Sigma-Aldrich

Methyl phenylacetate

≥98%, FCC, FG

Synonym(s):

Benzene acetic acid methyl ester, Methyl 2-phenylacetate, Phenyl-acetic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH3
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2733
Beilstein:
878795
EC Number:
Council of Europe no.:
2155
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.783
NACRES:
NA.21
Organoleptic:
honey; floral; spicy; sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.503 (lit.)

bp

218 °C (lit.)

density

1.066 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

honey; floral; spicy; sweet

SMILES string

COC(=O)Cc1ccccc1

InChI

1S/C9H10O2/c1-11-9(10)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

CRZQGDNQQAALAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl phenylacetate has been identified in the volatile fraction of beewax absolute oil, dried fruiting bodies of Boletus auripes, Chinese fermented black soybeans and peated malt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

211.1 °F - closed cup

Flash Point(C)

99.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the volatile components of peated malt. III. Identification of acidic and basic components
Deki M and Yoshimura M.
Chemical & Pharmaceutical Bulletin, 22(8), 1760-1764 (1974)
Analysis of Volatile Components of Boletus auripes Pk. by GC-MS.
Li W, et al.
Journal of Food Science and Technology, 8, 016-016 (2012)
Study on Volatile Constituents of Absolute Oil of Beeswax [J].
Sumin YXLJZ.
Flavour Fragrance Cosmetics, 4, 002-002 (2003)
Comparison of volatile flavor components in four kinds of Chinese fermented black soybeans (Glycine max (L.) Merrill) by HS-SPME-GC/MS and chemometrics.
Xiao ZB, et al.
Food Science and Technology Research, 17(4), 319-326 (2011)
Miriam Frida Karlsson et al.
Journal of agricultural and food chemistry, 57(13), 5903-5909 (2009-06-06)
The Guatemalan moth Tecia solanivora is an invasive pest of potato in Central and South America. The larvae infest potato tubers in the field as well as in storage facilities. The headspace of potato foliage and potato tubers was studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service