Skip to Content
Merck
All Photos(1)

Documents

18344

Supelco

Methyl benzoate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
1072099
EC Number:
MDL number:
UNSPSC Code:
85151701
eCl@ss:
39024501
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.68 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.516 (lit.)
n20/D 1.517

bp

198-199 °C (lit.)

mp

−12 °C (lit.)

solubility

H2O: soluble 2.1 g/L at 20 °C

density

1.088 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(=O)c1ccccc1

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl benzoate (MB) belongs to the class of benzenoid esters. It exists as a naturally occurring compound in a variety of plants including freshwater fern Salvinia molesta, Antirrhinum genus species, etc. MB extracted from the fermented apple juice is reported to possess toxicity against an invasive fruit fly, identified as Drosophila suzukii Matsumura.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Methyl benzoate may be used as an extraction solvent for extracting and preconcentrating benzimidazole fungicides in water samples prior to their determination by high-performance liquid chromatography with photodiode array detection (HPLC-PDA). It may also be used as an analytical reference standard for the quantification of the analyte in Rosa hybrida using gas chromatography-mass spectrometry technique (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A floral fragrance, methyl benzoate, is an efficient green pesticide
Feng Y and Zhang A
Scientific Reports, 7 (2017)
Alternative solvent?based methyl benzoate vortex?assisted dispersive liquid?liquid microextraction for the high?performance liquid chromatographic determination of benzimidazole fungicides in environmental water samples
Santaladchaiyakit Y and Srijarana S
Journal of Separation Science, 37(22), 3354-3361 (2014)
Genetic analysis of natural variation in antirrhinum scent profiles identifies benzoic acid carboxymethyl transferase as the major locus controlling methyl benzoate synthesis
Ruiz-Hernandez V, et al.
Frontiers in Plant Science, 8, 27-27 (2017)
Anna Klimek-Turek et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-04)
In this manuscript, the retention of aromatic hydrocarbons with polar groups has been compared for systems with various nonpolar columns of the types from C3 to C18 and different mobile phases composed of methanol, acetonitrile, or tetrahydrofuran as modifiers. The
María Manchado-Rojo et al.
The Plant journal : for cell and molecular biology, 72(2), 294-307 (2012-06-20)
The transcriptional network topology of B function in Antirrhinum, required for petal and stamen development, is thought to rely on initial activation of transcription of DEFICIENS (DEF) and GLOBOSA (GLO), followed by a positive autoregulatory loop maintaining gene expression levels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service