Skip to Content
Merck
All Photos(5)

Key Documents

W249166

Sigma-Aldrich

Furfuryl alcohol

greener alternative

natural, ≥95%, FG

Synonym(s):

2-(Hydroxymethyl)furan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
FEMA Number:
2491
Beilstein:
106291
EC Number:
Council of Europe no.:
2023
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.019
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 175.105

vapor density

3.4 (vs air)

vapor pressure

0.5 mmHg ( 20 °C)
1 mmHg ( 32 °C)
5.5 mmHg ( 55 °C)

Assay

≥95%

autoignition temp.

915 °F

expl. lim.

16.3 %

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.486 (lit.)

bp

170 °C (lit.)

mp

−29 °C (lit.)

density

1.135 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

bread; brown

storage temp.

2-8°C

SMILES string

OCc1ccco1

InChI

1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2

InChI key

XPFVYQJUAUNWIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural Occurrence: Burley tobacco, bread, cocoa, coffee, juniper berry oil, potato chips, roasted barley, whiskey
Furfuryl alcohol is a volatile flavor compound that occurs naturally in popcorn, rice cakes and sweet corn products.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application

Although this chemical doesn′t usually play a starring role in flavors, it will help to contribute brown, subtly smoky notes to Indonesian-type vanilla flavors, bacon and other smoked meats, lapsang souchong tea, whiskey, grain, coffee and barbecue flavors.

Biochem/physiol Actions

Taste at 20-40ppm

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nose, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Popcorn flavor: identification of volatile compounds.
Walradt JP, et al.
Journal of Agricultural and Food Chemistry, 18(5), 926-928 (1970)
Judith Michalski et al.
Perfumer & Flavorist, 39(2), 48-49 (2014)
Studies on flavor volatiles of some sweet corn products.
Buttery RG, et al.
Journal of Agricultural and Food Chemistry, 42(3), 791-795 (1994)
Annica Pilgård et al.
Environmental toxicology and chemistry, 29(9), 1918-1924 (2010-09-08)
The furfurylation process is an extensively investigated wood modification process. Furfuryl alcohol molecules penetrate into the wood cell wall and polymerize in situ. This results in a permanent swelling of the wood cell walls. It is unclear whether or not
Marco Minella et al.
Chemosphere, 90(2), 306-311 (2012-08-28)
Suspended particles in a system made up of Aldrich humic acids (HAs) in water account for about 13% of the total HA mass, 10-11% of the organic carbon and 9-11% of radiation extinction in the UVA region. Extinction would be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service