Skip to Content
Merck
All Photos(3)

Key Documents

W246665

Sigma-Aldrich

Eucalyptus oil

greener alternative

natural, ≥65%, product of Dutch Essential Oils, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2466
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

(Eucalyptus Globulus)

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU 1334/2008 & 178/2002
EU Regulation 1223/2009
FDA 21 CFR 117

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

≥65%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

limonene (sum of D, L and DL), Eucalyptus spp. leaf oil

greener alternative category

Organoleptic

camphoraceous; herbaceous

SMILES string

O1C2(CCC(C1(C)C)CC2)C

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3

InChI key

WEEGYLXZBRQIMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.8 °F - closed cup

Flash Point(C)

57.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service