Skip to Content
Merck
All Photos(2)

Key Documents

T82805

Sigma-Aldrich

Triphenylethylene

99%

Synonym(s):

1,1,2-Triphenylethene, 1,1,2-Triphenylethylene, 1,1′,1′′-(1-Ethenyl-2-ylidene)tris[benzene], 1,2-Diphenylethenylbenzene, Benzilidenediphenylmethane, Ethene-1,1,2-triyltribenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(C6H5)2
CAS Number:
Molecular Weight:
256.34
Beilstein:
1867462
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

69-71 °C (lit.)

SMILES string

c1ccc(cc1)\C=C(/c2ccccc2)c3ccccc3

InChI

1S/C20H16/c1-4-10-17(11-5-1)16-20(18-12-6-2-7-13-18)19-14-8-3-9-15-19/h1-16H

InChI key

MKYQPGPNVYRMHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triphenylethylene is an aromatic hydrocarbon, which can be used as a starting material to prepare 2,2,3-triphenyloxirane by asymmetric epoxidation reaction using fluorous chiral manganese complex as a catalyst. It is also used to prepare dihydro-4,5,5-triphenyl-2(3H)-furanone by reacting with acetic anhydride in the presence of MnO2 and NaOAc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Fang Li et al.
Journal of fluorescence, 21(5), 1969-1977 (2011-05-24)
New aggregation-induced emission (AIE) compounds derived from triphenylethylene were synthesized. The thermal, photophysical, electrochemical and aggregation-induced emissive properties were investigated. All the compounds had strong blue light emission capability and good thermal stability. Their maximum fluorescence emission wavelengths were between
[The search for active antitumor agents among triphenylethylene derivatives].
P V Sergeev et al.
Voprosy onkologii, 41(2), 49-49 (1995-01-01)
Effect of estrogenic and antiestrogenic triphenylethylene derivatives on progesterone and estrogen receptors levels of MCF-7 cells.
N Legros et al.
Biochemical pharmacology, 42(9), 1837-1841 (1991-10-09)
M Pons et al.
Journal of steroid biochemistry, 36(5), 391-397 (1990-08-14)
The relative binding affinities of a series of twelve para-hydroxylated triphenylethylenes (TPEs) for the estradiol receptor (ER) of calf uterus cytosol were measured by a competition method. The results obtained under equilibrium conditions support the hypothesis of the additivity of
Yuliya Dobrydneva et al.
Journal of cardiovascular pharmacology, 50(4), 380-390 (2007-12-01)
The anti-estrogenic drug tamoxifen, which is used therapeutically for treatment and prevention of breast cancer, can lead to the development of thrombosis. We found that tamoxifen rapidly increased intracellular free calcium [Ca2+]i in human platelets from both male and female

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service