Skip to Content
Merck
All Photos(2)

Key Documents

468045

Sigma-Aldrich

D-(+)-chiro-Inositol

95%

Synonym(s):

1,2,4/3,5,6-Hexahydroxycyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

optical activity

[α]23/D +60°, c = 1.2 in H2O

optical purity

ee: 99% (GLC)

mp

230 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@H]1[C@H](O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4-,5+,6+/m0/s1

InChI key

CDAISMWEOUEBRE-LKPKBOIGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The inositols and their phosphates have been used in the development of metabolically stable insulin mediators, inhibitors, and modulators of important metabolic functions such as glycolysis. Inositols are stable to degradative enzymes in vivo because they lack a hydrolytically labile glycosidic linkage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in the chemistry and biochemistry of inositol phosphates of biological interest.
B V Potter
Natural product reports, 7(1), 1-24 (1990-02-01)
Modern Methods of Monosaccharide Synthesis from Non-Carbohydrate Sources.
Tomas Hudlicky et al.
Chemical reviews, 96(3), 1195-1220 (1996-05-09)
Bellington, D.C.
Chemical Society Reviews, 18, 83-83 (1989)
M J Berridge et al.
Nature, 341(6239), 197-205 (1989-09-21)
Inositol 1,4,5-trisphosphate is a second messenger which regulates intracellular calcium both by mobilizing calcium from internal stores and, perhaps indirectly, by stimulating calcium entry. In these actions it may function with its phosphorylated metabolite, inositol 1,3,4,5-tetrakisphosphate. The subtlety of calcium
Hudlicky, T. et al.
Synthesis, 897-897 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service