Skip to Content
Merck
All Photos(1)

Key Documents

T81000

Sigma-Aldrich

Trioctylamine

98%

Synonym(s):

TOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]3N
CAS Number:
Molecular Weight:
353.67
Beilstein:
1210618
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.449 (lit.)

bp

164-168 °C/0.7 mmHg (lit.)
365-367 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN(CCCCCCCC)CCCCCCCC

InChI

1S/C24H51N/c1-4-7-10-13-16-19-22-25(23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3

InChI key

XTAZYLNFDRKIHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trioctylamine is used as an extractant for organic acids (such as TCA, succinic acid, and acetic acid), and precious metals.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Oral

Target Organs

Heart

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

334.4 °F - closed cup

Flash Point(C)

168 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongshuang Zhu et al.
Dalton transactions (Cambridge, England : 2003), 41(10), 2935-2940 (2012-01-24)
Cobalt phosphate-based organic-inorganic hybrid nanobelts were successfully synthesized in the presence of trioctylamine (TOA) via a solvothermal system. The inorganic cobalt phosphate layers and organic n-octylamine layers in this structure arranged alternately, both of which were parallel to the axial
High pressure liquid chromatographic determination of tartrazine in rice milk following ion-pair extraction with tri-n-octylamine.
M L Puttemans et al.
Journal - Association of Official Analytical Chemists, 66(3), 670-672 (1983-05-01)
Yun Suk Huh et al.
Biotechnology letters, 26(20), 1581-1584 (2004-12-18)
Acetic acid is by-product from fermentation processes for producing succinic acid using Mannheimia succiniciproducens . To obtain pure succinic acid from the final fermentation broth, acetic acid was selectively removed based on the different extractability of succinic acid and acetic
C N Lin et al.
Biotechnology and bioengineering, 69(1), 31-38 (2000-05-23)
A lipase-catalyzed enantioselective hydrolysis process under conditions of continuous in situ racemization of substrate with trioctylamine as the catalyst was developed for the production of (S)-suprofen from (R,S)-suprofen 2,2,2-trifluoroethyl thioester in isooctane. A detailed investigation of trioctylamine concentration on the
M L Puttemans et al.
Journal - Association of Official Analytical Chemists, 67(5), 880-885 (1984-09-01)
Synthetic dyes, benzoic acid, sorbic acid, and saccharin are extracted simultaneously from soft drinks with 0.01M tri-n-octylamine at pH = 5.5 and are back-extracted to an aqueous phase with 0.1M sodium perchlorate. The perchlorate solution is injected directly into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service