Skip to Content
Merck
All Photos(1)

Key Documents

N16754

Sigma-Aldrich

2-Nitrofluorene

98%

Synonym(s):

2-Nitro-9H-fluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO2
CAS Number:
Molecular Weight:
211.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder

mp

156-158 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C13H9NO2/c15-14(16)11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7H2

InChI key

XFOHWECQTFIEIX-UHFFFAOYSA-N

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Hirayama et al.
Carcinogenesis, 21(11), 2105-2111 (2000-11-04)
Although the intestinal flora is believed to have a critical role in carcinogenesis, little is known about the role of the human intestinal flora on the effects of mutagens in vivo. The aim of the present study was to address
W A Kappers et al.
Mutation research, 466(2), 143-159 (2000-03-23)
Three different in vitro mutation assays were used to investigate the involvement of cytochrome P450 enzymes in the activation of the nitro-polycyclic aromatic hydrocarbons (nitroPAHs) 1-nitropyrene and 2-nitrofluorene and their reduced metabolites amino-polycyclic aromatic hydrocarbons (aminoPAHs) 1-aminopyrene and 2-aminofluorene. Mutagenicity
Ray A Matulka et al.
Frontiers in veterinary science, 6, 477-477 (2020-01-24)
Chickens exposed to antigens produce IgY antibodies, similar in structure to mammalian IgG. Hens exposed with an allergen produced by cats (Fel d 1) results in production of anti-Fel d 1 specific IgY (AFD1), which is naturally concentrated in egg
Valentina Nanni et al.
Foods (Basel, Switzerland), 9(10) (2020-10-22)
Plant secondary metabolites possess chemopreventive and antineoplastic properties, but the lack of information about their exact mechanism of action in mammalian cells hinders the translation of these compounds in suitable therapies. In light of this, firstly, Origanum vulgare L. hydroalcoholic
Zhonghan Hu et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(4), 831-836 (2006-01-19)
In this work, we investigate the slow dynamics of 1-butyl-3-methylimidazolium hexafluorophosphate, a very popular room-temperature ionic solvent. Our study predicts the existence of heterogeneity in the liquid and shows that this heterogeneity is the underlying microscopic cause for the recently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service