M12002
1-Methoxy-1,3-butadiene
95%
Synonym(s):
1,3-Butadienyl methyl ether, 1-Methoxy-1,3-butadiene, 1-Methoxybutadiene, 4-Methoxy-1,3-butadiene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH2=CHCH=CHOCH3
CAS Number:
Molecular Weight:
84.12
Beilstein:
1420823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.464 (lit.)
bp
91 °C (lit.)
density
0.83 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CO\C=C\C=C
InChI
1S/C5H8O/c1-3-4-5-6-2/h3-5H,1H2,2H3/b5-4+
InChI key
KOCUMXQOUWPSLK-SNAWJCMRSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
21.2 °F - closed cup
Flash Point(C)
-6 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service