Skip to Content
Merck
All Photos(1)

Key Documents

JKA8008

Sigma-Aldrich

8arm-PEG10K

average MN 10,000, NH2, hexaclycerol core, HCl Salt, carboxyl reactive

Synonym(s):

8arm-PEG-NH2, hexaglycerol core, 8arm-PEG-NH2, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
R(O(CH2CH2O)nCH2CH2NH2)8
UNSPSC Code:
12162002
NACRES:
NA.23

product name

8arm-PEG10K-NH2, hexaglycerol core, HCl Salt, average Mn 10,000

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: carboxyl reactive

polymer architecture

shape: 8-arm
functionality: homofunctional

Looking for similar products? Visit Product Comparison Guide

General description

8arm PEG Amine. Crosslinks into hydrogels. Amine group reacts with carboxylic group (-COOH) or other amine reactive chemical groups.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frith, J.E., et al.
Biomaterials, 35(4) (2014)
Giorgi, M.E., et al.
Glycobiology, 22(10) (2012)
Jeon, O., et al.
Acta Biomaterialia, 10(1) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service