Skip to Content
Merck
All Photos(1)

Key Documents

JKP2003R

Sigma-Aldrich

4arm-PEG10K

average Mn 10,000

Synonym(s):

4arm-PEG, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2O(CH2CH2O)nH)4
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 10,000

storage temp.

−20°C

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Farooque, T.M., et al.
Biomaterials, 35(9) (2014)
Xu, J., E. Feng, et al.
Journal of the American Chemical Society, 136(11) (2014)
Azagarsamy, M.A., et al.
ACS Macro Letters, 3(6) (2014)
Tokuda, E.Y., et al.
Biomaterials, 35(14) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service