Skip to Content
Merck
All Photos(3)

Key Documents

I3408

Sigma-Aldrich

Indole

≥99%

Synonym(s):

1H-Benzo[b]pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
Beilstein:
107693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

bp

253-254 °C (lit.)

mp

51-54 °C (lit.)

SMILES string

c1ccc2[nH]ccc2c1

InChI

1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H

InChI key

SIKJAQJRHWYJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Indole is a nitrogen-containing heterocycle used in the total synthesis of compounds such as goniomitine, (−)-isatisine A, and (±)-aspidospermidine.
  • It is used as an electron donor moiety in synthesizing dyes for organic photovoltaics.
  • It can also be used in the preparation of indole based conjugated small molecules for nonlinear optics applications.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total Synthesis of (−)-Isatisine A
Zhang X, et al.
Angewandte Chemie (International ed. in English), 50(27), 6164-6166 (2011)
Synthesis of double D-A branched organic dyes employing indole and phenoxazine as donors for efficient DSSCs
Hong Y, et al.
Tetrahedron, 70(36), 6296-6302 (2014)
Synthesis and nonlinear optical properties of novel conjugated small molecules based on indole donor
Liu J, et al.
Journal of Molecular Structure, 1165(30), 223-227 (2018)
Impact of Thermal Annealing on Organic Photovoltaic Cells Using Regioisomeric Donor-Acceptor-Acceptor Molecules
Zhang T, et al.
ACS Applied Materials & Interfaces, 9(30), 25418-25425 (2017)
Catalytic selective cyclizations of aminocyclopropanes: formal synthesis of aspidospermidine and total synthesis of goniomitine
De Simone F, et al.
Angewandte Chemie (International ed. in English), 122(33), 5903-5906 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service