Skip to Content
Merck
All Photos(1)

Key Documents

1.11350

Millipore

Bactident Kovac′s Indole reagent

Available in a practical dropper bottle, for Escherichia coli

Synonym(s):

4-(Dimethylamino)benzaldehyde solution, Indole reagent according to Kovac, Indole reagent, Indole test reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.84

Quality Level

description

Bactident™ E. coli 1.13303

form

liquid (ready-to-use)

shelf life

210 days

feature

ready-to-use

composition

(n-Butanol; Hydrochloric acid; 4 dimethylaminobenzaldehyde)

packaging

pkg of (1 x 30 ml, Plastic bottle)

technique(s)

microbe id | specific enzyme detection: suitable

pH

<1 (20 °C in H2O)

transition temp

flash point 36 °C

density

0.92 g/cm3 at 20 °C

application(s)

environmental
food and beverages

storage temp.

2-8°C

suitability

Enterobacter spp.
Escherichia coli

General description

Kovac′s reagent is a biochemical reagent composed of n-Butanol, hydrochloric acid, and 4-dimethylaminobenzaldehyde. It is used for the detection of indole producing microorganisms. Tryptophan is abundant in trypticalle digested peptone. Some microorganisms can cleave tryptophan to give pyruvic acid, ammonia, and indole. Indole then reacts with 4-dimethylaminobenzaldehyde to form a dark red dye. As tryptophan also gives a color reaction with 4 dimethylaminobenzaldehyde, it must be separated from the indole. This is achieved by selectively extracting indole with butanol.

Application

Bactident Kovac′s Indole reagent is used for the detection of indole producing microorganisms in food and beverages, and environmental samples. This is suitable for the identification of E. coli.

Linkage

Replaces: 1.11350.0007

Analysis Note

Indole formation
- Escherichia coli ATCC 25922 (WDCM 00013):
+
- Proteus vulgaris ATCC 8427: +
- Enterobacter aerogenes ATCC 13048: -
Incubation: 24 hrs.; 35 °C; aerobic.
Reference: DEV-Tryptophan broth.

Legal Information

BACTIDENT is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service