Skip to Content
Merck
All Photos(3)

Key Documents

H7408

Sigma-Aldrich

Hexadecylamine

technical grade, 90%

Synonym(s):

1-Aminohexadecane, Cetylamine, HAD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15NH2
CAS Number:
Molecular Weight:
241.46
Beilstein:
1634065
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor pressure

<1 mmHg ( 20 °C)

Assay

90%

bp

330 °C (lit.)

mp

43-45 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCN

InChI

1S/C16H35N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-17H2,1H3

InChI key

FJLUATLTXUNBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hexadecylamine, also known as cetylamine, is commonly used in amidation reactions to introduce amide functional groups. It serves as a key building block for surfactants and plays a significant role in catalysis and ligand synthesis.Additionally, hexadecylamine is employed to modify functional groups in various organic compounds.

Application

  • Interfacial structuring of liquids: Hexadecylamine is explored for its role in the solvent-assisted interfacial association of oppositely charged polyelectrolytes and amphiphiles, offering potential advancements in the structuring of liquids for various industrial and scientific applications (Bak et al., 2023).
  • Efficient light-to-thermal conversion: Additionally, hexadecylamine is used in ZnO nanorods loading with fatty amine as composite phase change materials (PCMs) device, enhancing light-to-thermal and electro-to-thermal energy conversion, applicable in energy storage technologies (Cao et al., 2023).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Gastrointestinal tract,Liver,Immune system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

285.8 °F - closed cup

Flash Point(C)

141 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Hexadecylamine as both reducing agent and stabilizer to synthesize Au and Ag nanoparticles and their SERS application
Hou et al.
Journal of Nanoparticle Research, 13, 1929-1936 (2011)
Study on Syntheses and Properties of Sodium Monoitaconamic Carboxylate Surfactants With Amide Group
Jun et al.
IOP Conference Series: Materials Science and Engineering, 392, 052024-052024 (2018)
Mesoporous titania nanocrystals prepared using hexadecylamine surfactant template: crystallization progress monitoring, morphological characterization and application in dye-sensitized solar cells
Alexaki et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 124, 52-58 (2009)
Sheng-Chih Lin et al.
Nature communications, 11(1), 3525-3525 (2020-07-16)
Copper electrocatalysts have been shown to selectively reduce carbon dioxide to hydrocarbons. Nevertheless, the absence of a systematic study based on time-resolved spectroscopy renders the functional agent-either metallic or oxidative Copper-for the selectivity still undecidable. Herein, we develop an operando
Shape-controlled synthesis of copper nanocrystals in an aqueous solution with glucose as a reducing agent and hexadecylamine as a capping agent.
Mingshang Jin et al.
Angewandte Chemie (International ed. in English), 50(45), 10560-10564 (2011-09-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service