Skip to Content
Merck
All Photos(2)

Key Documents

GF45743698

Erbium

foil, 25x25mm, thickness 0.125mm, as rolled, 99%

Synonym(s):

Erbium, ER000215

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Er
CAS Number:
Molecular Weight:
167.26
MDL number:
UNSPSC Code:
12141603
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 457-436-98

resistivity

86 μΩ-cm, 20°C

size × thickness

25 x 25 mm × 0.125 mm

bp

2868 °C (lit.)

mp

1529 °C (lit.)

density

9.062 g/mL at 25 °C (lit.)

SMILES string

[Er]

InChI

1S/Er

InChI key

UYAHIZSMUZPPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald J Caniglia
Facial plastic surgery clinics of North America, 12(3), 373-377 (2004-07-21)
The Erbium:YAG laser is a useful flexible tool for the surgeon interested in providing overall skin rejuvenation. With its unique wavelength, greater absorption, and less thermal injury, it can offer more precise ablation with greater safety and shorter healing times.
Isao Ishikawa et al.
Journal of periodontal research, 39(4), 275-285 (2004-06-23)
Since lasers were introduced for the treatment of oral diseases, there has been considerable advancement in technology. As a result, numerous laser systems are currently available for oral use. Neodymium:Yttrium-Aluminum:Garnet (Nd:YAG), carbon dioxide (CO(2)) laser and the semiconductor Diode lasers
Bo Li et al.
Journal of nanoscience and nanotechnology, 14(6), 4191-4195 (2014-04-18)
We have synthesized Er2S3/ZnS core/shell QDs by employing ErSt3, ZnSt2, and sulfur as precursors via a hot solution phase chemistry using a nucleation-doping strategy. X-ray diffraction (XRD), transmission electron microscope (TEM) and photoluminescence (PL) spectra were used to characterize the
D M Clarkson
Dental update, 28(6), 298-302 (2001-08-31)
This article reviews aspects of the probable mechanisms used by erbium dental lasers for cutting dentine and enamel, describes key issues of the risk of temperature elevation and speed of cutting relative to conventional techniques and looks at issues concerned
G N Marcells et al.
The Journal of otolaryngology, 29(2), 78-82 (2000-05-20)
There are two lasers for resurfacing. The erbium:YAG laser and the CO2 laser both have different physical and physiologic differences when striking the skin. Therefore, each laser gives a different result. This article reviews the use of the Sharplan Silktouch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service