Skip to Content
Merck
All Photos(2)

Key Documents

GF36861701

Europium

wire reel, 50mm, diameter 1.0mm, hard, 99.95%

Synonym(s):

Europium, EU005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Eu
CAS Number:
Molecular Weight:
151.96
MDL number:
UNSPSC Code:
12141604
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

wire

manufacturer/tradename

Goodfellow 368-617-01

resistivity

81 μΩ-cm, 20°C

L × diam.

50 mm × 1.0 mm

bp

1527 °C (lit.)

mp

822 °C (lit.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

[Eu]

InChI

1S/Eu

InChI key

OGPBJKLSAFTDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Line Storm et al.
Cancer immunology, immunotherapy : CII, 64(1), 41-50 (2014-09-28)
Colorectal cancer (CRC) is a leading cause of cancer death worldwide. Lack of symptoms results in late detection and increased mortality. Inflammation, including complement activation, plays an important role in tumorigenesis. The concentrations of nine proteins of the lectin pathway
M L P Reddy et al.
Dalton transactions (Cambridge, England : 2003), 42(43), 15249-15262 (2013-10-01)
Visible-light sensitized luminescent europium(III) molecular materials are of considerable importance because their outstanding photophysical properties make them well suited as labels in fluorescence-based bioassays and low-voltage driven pure red-emitters in optoelectronic technology. One challenge in this field is development of
Satoshi Sakamoto et al.
Clinical chemistry, 60(4), 610-620 (2014-01-18)
Accurate detection and monitoring of disease-related biomarkers is important in understanding pathophysiology. We devised a rapid immunoreaction system that uses submicrometer polymer-coated fluorescent ferrite (FF) beads containing both ferrites (magnetic iron oxide) and fluorescent europium complexes. FF beads were prepared
Tsung-Sheng Wu et al.
Molecular endocrinology (Baltimore, Md.), 28(7), 1026-1038 (2014-06-04)
SHBG transports and regulates the activities of androgens and estrogens. Several single nucleotide polymorphisms in the human SHBG gene have been linked to sex steroid-dependent diseases, including those associated with the metabolic syndrome. The N-terminal laminin G-like domain of SHBG
Soo-Kyung Kim et al.
ChemMedChem, 9(8), 1732-1743 (2014-07-06)
Human Urotensin-II (U-II) is the most potent mammalian vasoconstrictor known.1 Thus, a U-II antagonist would be of therapeutic value in a number of cardiovascular disorders.2 Here, we describe our work on the prediction of the structure of the human U-II

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service