Skip to Content
Merck
All Photos(1)

Documents

D35457

Sigma-Aldrich

N,N-Dibenzylhydroxylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2NOH
CAS Number:
Molecular Weight:
213.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

125-128 °C (lit.)

SMILES string

ON(Cc1ccccc1)Cc2ccccc2

InChI

1S/C14H15NO/c16-15(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10,16H,11-12H2

InChI key

GXELTROTKVKZBQ-UHFFFAOYSA-N

Application

N,N-Dibenzylhydroxylamine, upon oxidation, yields N-benzyl-α-phenylnitrone, which can undergo cycloaddition reaction with suitable dipolarophiles. It can be used to synthesize N,N,O-trisubstituted hydroxylamines and arylamines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of N,N,O-Trisubstituted Hydroxylamines by Stepwise Reduction and Substitution of O-Acyl N, N-Disubstituted Hydroxylamines.
Dhanju S and Crich D
Organic Letters, 18(8), 1820-1823 (2016)
Mechanistic studies lead to dramatically improved reaction conditions for the Cu-catalyzed asymmetric hydroamination of olefins.
Bandar JS, et al.
Journal of the American Chemical Society, 137(46), 14812-14818 (2015)
Synthesis of Arylamines via Aminium Radicals.
Svejstrup TD, et al.
Angewandte Chemie (International Edition in English), 56(47), 14948-14952 (2017)
Recent developments in nitrone chemistry: some novel transformations.
Yang J
Synlett, 23(16), 2293-2297 (2012)
Chemoselective Ligation of Peptide Phenyl Esters with N?Terminal Cysteines.
Fang GM, et al.
Chembiochem, 11(8), 1061-1065 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service