Skip to Content
Merck
All Photos(3)

Documents

D193402

Sigma-Aldrich

3,5-Dinitroaniline

≥96.5% purity, powder, crystals or chunks

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NH2
CAS Number:
Molecular Weight:
183.12
Beilstein:
648811
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

3,5-Dinitroaniline, ≥96.5% (GC)

Assay

≥96.5% (GC)

form

powder, crystals or chunks

color

faint yellow to dark yellow, and Faint Brown to Dark Brown

mp

160-162 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H5N3O4/c7-4-1-5(8(10)11)3-6(2-4)9(12)13/h1-3H,7H2

InChI key

MPBZUKLDHPOCLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ru Chen et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-22)
Functional polymeric materials composed solely of lignin preparations appeared only very recently. A gradual paradigm shift spanning 56 years has revealed how lignin-lignin blends can upgrade the performance of 100 wt% lignin-based plastics. The view, first espoused in 1960, that
Hong-Bin Luo et al.
ACS nano, 13(7), 8417-8424 (2019-07-04)
Organic color center-tailored semiconducting single-walled carbon nanotubes are a rising family of synthetic quantum emitters that display bright defect photoluminescence molecularly tunable for imaging, sensing, and quantum information processing. A major advance in this area would be the development of
Rachel C Irlam et al.
Journal of chromatography. A, 1629, 461506-461506 (2020-09-01)
The development of a new, lower cost method for trace explosives recovery from complex samples is presented using miniaturised, click-together and leak-free 3D-printed solid phase extraction (SPE) blocks. For the first time, a large selection of ten commercially available 3D
Daniel Bouša et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(49), 17728-17738 (2015-10-24)
In the last decade, graphene and graphene derivatives have become some of the most intensively studied materials. Tuning of the electronic and electrochemical properties of graphene is of paramount importance. In this study, six diazonium-modified graphenes containing different functional groups
Helena Rapp-Wright et al.
Journal of hazardous materials, 329, 11-21 (2017-01-26)
The first comprehensive assessment of 34 solid phase extraction sorbents is presented for organic explosive residues in wastewater prior to analysis with liquid chromatography-high resolution accurate mass spectrometry (LC-HRMS). A total of 18 explosives were selected including nitramines, nitrate esters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service