Skip to Content
Merck
All Photos(2)

Documents

D193011

Sigma-Aldrich

2,4-Dinitroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NH2
CAS Number:
Molecular Weight:
183.12
Beilstein:
982999
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

contains

≤15% water

mp

176-178 °C (lit.)

SMILES string

Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H5N3O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H,7H2

InChI key

LXQOQPGNCGEELI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

435.2 °F - closed cup

Flash Point(C)

224 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojia Huang et al.
Journal of chromatography. A, 1216(20), 4354-4360 (2009-04-03)
A simple and sensitive method for the determination of polar aromatic amines (PAAs) was developed using stir bar sorptive extraction (SBSE) coupling to high-performance liquid chromatography. A hydrophilic poly(vinylimidazole-divinylbenzene) (VIDB) monolithic material was prepared and acted as SBSE coating. The
B Striepen et al.
The Journal of cell biology, 151(7), 1423-1434 (2001-01-03)
Apicomplexan parasites harbor a single nonphotosynthetic plastid, the apicoplast, which is essential for parasite survival. Exploiting Toxoplasma gondii as an accessible system for cell biological analysis and molecular genetic manipulation, we have studied how these parasites ensure that the plastid
[Establishment of the MPEL for 2,4-dinitroaniline in water by an experimental computational method].
N Ia Mikhaĭlovskiĭ et al.
Gigiena i sanitariia, (10)(10), 15-18 (1982-10-01)
S P Ozheredov et al.
TSitologiia i genetika, 43(5), 3-13 (2010-05-13)
The results of Allium-test screening of new 2,4- and 2,6-dinitroaniline derivates on antimitotic activity and phytotoxicity are presented in the work. It is revealed that all studied compounds which are derivates of 2,4-dinitroaniline, 2,6-dinitro-(4-fluoromethyl)-aniline as well as (methylsulfonyl) nitrobenzol, can
P Gettins et al.
The Biochemical journal, 197(1), 119-125 (1981-07-01)
Pre-resonance Raman spectroscopy was used to study the interactions of the nitro groups of dinitrophenyl haptens with three dinitrophenyl-binding antibody fragments: M315 Fv, M460 Fab' and X25 Fab'. The observed changes in frequency of modes associated with the nitro moieties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service