Skip to Content
Merck
All Photos(1)

Key Documents

D161292

Sigma-Aldrich

2-Amino-6-methylheptane

99%

Synonym(s):

1,5-Dimethylhexylamine, 6-Methyl-2-heptylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3CH(CH3)NH2
CAS Number:
Molecular Weight:
129.24
Beilstein:
1209250
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.422 (lit.)

bp

154-156 °C (lit.)

density

0.767 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCC(C)N

InChI

1S/C8H19N/c1-7(2)5-4-6-8(3)9/h7-8H,4-6,9H2,1-3H3

InChI key

QNIVIMYXGGFTAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Amino-6-methylheptane can be used as:
  • A building block for the synthesis of n-type photoconductor, N,N′-bis(1,5-dimethylhexyl)-3,4:9,10-perylenebis (dicarbox-imide) (PDHEP).
  • A reagent in the synthesis of substituted pyrrolidones by reductive amination of levulinic acid using supported platinum catalyst.
  • A nucleophilic amine in the synthesis of unsymmetrical sulfamide by reacting with sulfamic acid salts and triphenylphosphine ditriflate.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted synthesis of phthalocyanine-porphyrin complex and its photoelectric conversion properties
Liu MO and Hu AT
Journal of Organometallic Chemistry, 689(15), 2450-2455 (2004)
Heterogeneous Pt catalysts for reductive amination of levulinic acid to pyrrolidones
Touchy AS, et al.
ACS Catalysis, 4(9), 3045-3050 (2014)
Efficient synthesis of unsymmetrical sulfamides from sulfamic acid salts by activation with triphenylphosphine ditriflate
Shehata MF, et al.
Tetrahedron, 75(24), 3186-3194 (2019)
Pieter A Cohen et al.
Clinical toxicology (Philadelphia, Pa.), 56(6), 421-426 (2017-11-09)
The United States Food and Drug Administration banned the stimulant 1,3-dimethylamylamine (1,3-DMAA) from dietary supplements and warned consumers that the stimulant can pose cardiovascular risks ranging from high blood pressure to heart attacks. We designed our study to determine if

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service