Skip to Content
Merck
All Photos(3)

Key Documents

D115606

Sigma-Aldrich

1,5-Dihydroxynaphthalene

97%

Synonym(s):

1,5-Naphthalenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(OH)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
2044951
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

259-261 °C (dec.) (lit.)

SMILES string

Oc1cccc2c(O)cccc12

InChI

1S/C10H8O2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6,11-12H

InChI key

BOKGTLAJQHTOKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Baumann et al.
Prostaglandins, 20(4), 627-639 (1980-10-01)
Until now only few data have been reported on biochemically explicable pharmacological effects of flavonoid structures. When tested against arachidonic acid metabolism many flavonoids were found to be effective against the lipoxygenase and cyclo-oxygenase pathways. Some flavonoids were predominant inhibitors
Shin-ya Takizawa et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(6), 895-903 (2011-02-08)
Photooxidation reactions of 1,5-dihydroxynaphthalene (DHN) have been carried out in the presence of cyclometalated neutral and cationic iridium (Ir) complexes 1-6 as singlet oxygen ((1)O(2)) sensitizers in order to investigate the (1)O(2) generation quantum yield and photosensitizing durability of the
S Gal et al.
Chemistry and physics of lipids, 150(2), 186-203 (2007-09-29)
Copper-induced peroxidation of liposomal palmitoyllinoleoyl-phosphatidylcholine (PLPC) is inhibited by alpha-tocopherol at micromolar concentrations. In our previous study we found that when the liposomes contain phosphatidylserine (PS), nanomolar concentrations of Toc were sufficient to inhibit peroxidation. In an attempt to gain
Interactions of sulfhydryl agents and soybean lipoxygenase inhibitors.
I Knippel et al.
Biochemical pharmacology, 30(12), 1677-1684 (1981-06-15)
Yifan Liu et al.
Chemical communications (Cambridge, England), 48(31), 3751-3753 (2012-03-09)
Visible light-harvesting perylenebisimide (PBI)-C(60) dyads were prepared as organic triplet photosensitizers for photooxidation of 1,5-dihydroxynaphthalene and the efficiency of the dyads is 6-fold of the conventional Ir(III) complex triplet photosensitizer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service