Skip to Content
Merck
All Photos(1)

Key Documents

70430

Sigma-Aldrich

1,4-Dihydroxynaphthalene

technical, ≥90% (HPLC)

Synonym(s):

1,4-Naphthalenediol, p-Naphthohydroquinone (α)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
Beilstein:
1307689
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (HPLC)

form

crystals

mp

191-192 °C (dec.)

SMILES string

Oc1ccc(O)c2ccccc12

InChI

1S/C10H8O2/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6,11-12H

InChI key

PCILLCXFKWDRMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Teresa Molina et al.
The Journal of organic chemistry, 74(24), 9573-9575 (2009-11-27)
The NHC-catalyzed conjugate hydroacylation of 1,4-naphthoquinones allows for the synthesis of monoacylated 1,4-dihydroxynaphthalene derivatives. These targets, difficult to prepare selectively by standard protocols, represent important intermediates in the elaboration of highly substituted 1,4-naphthoquinone derivatives, which constitute relevant pharmaceutical scaffolds. High
R Munday
Free radical biology & medicine, 22(4), 689-695 (1997-01-01)
At neutral pH, 1,4-naphthohydroquinone and 2-methyl-1,4-naphthohydroquinone readily autoxidize to the corresponding quinones. In an unpurified phosphate buffer, the autoxidation of both substances proceeded in a linear fashion after a brief lag phase. Addition of a chelating agent or purification of
M Matsumoto et al.
Biological & pharmaceutical bulletin, 24(3), 236-241 (2001-03-21)
The proteases encoded by herpesviruses including herpes simplex virus type 1 (HSV-1) and human cytomegalovirus (HCMV) are attractive targets for antiviral drug development because of their important roles in viral replication. We randomly screened a chemical compound library for inhibitory
Aurora Molinari et al.
Archiv der Pharmazie, 342(10), 591-599 (2009-09-16)
Several new 6-(3-pyrazolylpropyl) derivatives of 1,4-naphthohydroquinone-1,4-diacetate (NHQ-DA) have been prepared by chemical modifications of the Diels-Alder adduct of alpha-myrcene and 1,4-benzoquinone. All these new compounds and precursors have been evaluated in vitro for their cytotoxicity against cultured human cancer cells
L S Tsuruda et al.
Archives of toxicology, 69(6), 362-367 (1995-01-01)
Naphthalene (NA) is metabolically activated to the reactive intermediates, naphthalene oxide (NO) and naphthoquinones. To investigate the role of circulating reactive metabolites in NA toxicity, the half-life of NO was examined. The in vitro half-life of NO in both whole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service