Skip to Content
Merck
All Photos(1)

Key Documents

CDS019982

Sigma-Aldrich

4H-1,2,4-Triazol-3-amine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N4
Molecular Weight:
84.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

form

solid

SMILES string

Nc1nnc[nH]1

InChI

1S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6)

InChI key

KLSJWNVTNUYHDU-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuchen Long et al.
The Plant cell, 27(4), 1185-1199 (2015-04-02)
Plant cells cannot rearrange their positions; therefore, sharp tissue boundaries must be accurately programmed. Movement of the cell fate regulator SHORT-ROOT from the stele to the ground tissue has been associated with transferring positional information across tissue boundaries. The zinc
Guangheng Wu et al.
The Plant cell, 27(3), 857-873 (2015-03-10)
Obligate biotrophs, such as the powdery mildew pathogens, deliver effectors to the host cell and obtain nutrients from the infection site. The interface between the plant host and the biotrophic pathogen thus represents a major battleground for plant-pathogen interactions. Increasing
B M Essa et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 76, 102-109 (2015-05-10)
Lactoperoxidase (LPO) inhibitors are very selective for solid tumor due to their high binding affinity to the LPO enzyme. A computational study was used to select top-ranked LPO inhibitor (alone and in complex with (99m)Tc) with high in silico affinity.
Toni R Bartling et al.
Free radical biology & medicine, 74, 99-107 (2014-06-29)
Aberrant matrix metalloproteinase-1 (MMP-1) expression contributes to the pathogenesis of many degenerative disease processes that are associated with increased oxidative damage or stress. We and others have established that shifts in steady-state H2O2 production resulting from enforced antioxidant gene expression
Girdhar K Pandey et al.
Plant physiology, 169(1), 780-792 (2015-07-23)
The role of calcium-mediated signaling has been extensively studied in plant responses to abiotic stress signals. Calcineurin B-like proteins (CBLs) and CBL-interacting protein kinases (CIPKs) constitute a complex signaling network acting in diverse plant stress responses. Osmotic stress imposed by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service