BR4760141
BRAND® Titrette® digital bottle-top burette
capacity 10 mL, without interface RS 232
Synonym(s):
Bottle-top Burette, Bottle-top Dispenser, Burette, Dispenser, Titrette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
Recommended Products
feature
without interface RS 232
manufacturer/tradename
BRAND 4760141
volume measuring range
10 mL
capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Titrette® is the first bottle-top burette to meet class A error limits for glass burettes. In addition, it stands out for easy titration, a compact design, excellent precision, an exchangeable dosing unit, and an optional PC interface. You can use the Titrette® to titrate quickly and reliably, even with restricted space and independent of available power connections - in the laboratory, in production, or in the field.
Application
Titrating
Features and Benefits
Overview:
Limitations on use:
- Simple: Titration within Class A error limits directly from the bottle
- Easy: Compact and robust
- Efficient: Fast filling, precise dispensing
- Separate buttons for on/off, pause, and CLEAR to delete the display and select function
- Non-slip hand wheels and a smooth-running precision gear set for fast or drip titration make handling reliable.
Limitations on use:
- Operating temperature from +15 °C to +40 °C (from 59 °F to 104 °F) of instrument and reagent
- Vapor pressure up to 500 mbar
- Viscosity up to 500 mm2 /s
- Altitude: max. 3,000 m above sea level
- Relative humidity: 20 % to 90%
Components
Items supplied:
With PC Interface, also includes:
- Titrette bottle-top burette
- DE-M marking
- Performance certificate
- Telescoping filling tube (170 - 330 mm)
- Recirculation tube
- 2 micro batteries (AAA/UM4/LR03)
- 3 bottle adapters PP (GL 45/32, GL 45/S 40 und GL 32/NS 29/32)
- 2 colored light shield inspection windows
With PC Interface, also includes:
- Connector cable (Sub-D plug connector, 9-pin)
- CD with driver software
- Communication protocol
- Example application
- Operating manual
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service