BR4600330
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 0.5-5 mL, without recirculation valve
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
incremental 0.02 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4600330
volume range
0.5-5 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)