BR4600130
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.5-5 mL, without recirculation valve
Synonym(s):
bottle top dispensers
All Photos(1)
About This Item
Recommended Products
feature
incremental 0.1 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4600130
volume range
0.5-5 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
Related Categories
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)