All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C9H6BrN
CAS Number:
Molecular Weight:
208.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
crystals
bp
280-285 °C (lit.)
mp
40-43 °C (lit.)
storage temp.
2-8°C
SMILES string
Brc1cncc2ccccc12
InChI
1S/C9H6BrN/c10-9-6-11-5-7-3-1-2-4-8(7)9/h1-6H
InChI key
SCRBSGZBTHKAHU-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service