Skip to Content
Merck
All Photos(1)

Key Documents

696439

Sigma-Aldrich

(S)-Ethyl piperidine-3-carboxylate

97%

Synonym(s):

(S)-3-Piperidinecarboxylic acid ethyl ester, (S)-Ethyl nipecotate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.471

density

1.043 g/mL at 25 °C

functional group

ester

SMILES string

CCOC(=O)[C@H]1CCCNC1

InChI

1S/C8H15NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h7,9H,2-6H2,1H3/t7-/m0/s1

InChI key

XIWBSOUNZWSFKU-ZETCQYMHSA-N

Application

Reactant for synthesis of:
  • DPP-4 inhibitors
  • JAK2 inhibitors
  • Serotonin and noradrenaline reuptake inhibitors
  • Human tryptase inhibitors
  • Anti-thrombotic nicecotamides

Reactive for oxidative C-arylation of free (NH)-heterocycles via direct (sp3) C-H bond functionalization

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service