Skip to Content
Merck
All Photos(2)

Key Documents

A70402

Sigma-Aldrich

2-Amino-4-nitrophenol

96%

Synonym(s):

2-Hydroxy-5-nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
776533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

form

solid

mp

140-143 °C (lit.)

SMILES string

Nc1cc(ccc1O)[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-5-3-4(8(10)11)1-2-6(5)9/h1-3,9H,7H2

InChI key

VLZVIIYRNMWPSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Eremin
Prikladnaia biokhimiia i mikrobiologiia, 36(4), 449-457 (2000-09-20)
Characteristics of polydisulfides of gallic acid (PDSG), 2-amino-4-nitrophenol (PDSANP), and biuret (PDSB) depending on the composition of the aqueous medium were studied. In contrast to PDSANP and PDSB, there was oxidation of PDSG with accumulation of products of polydisulfide transformation
2-Amino-4-nitrophenol.
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 167-176 (1993-01-01)
T A Robert et al.
Journal of chromatography, 344, 177-186 (1985-11-08)
Eleven groups of six ICR mice were dosed orally with 22.5 mg/kg 2,4-dinitrophenol. Groups were sacrificed at 0, 0.5, 1, 2, 4, 6, 9, 12, 24, 48, and 96 h post-treatment and plasma was collected for analysis of dinitrophenol, 2-amino-4-nitrophenol
D I Metelitza et al.
Journal of inorganic biochemistry, 98(1), 1-9 (2003-12-09)
The steady-state kinetics of the horseradish peroxidase (HRP)-catalyzed oxidation of 3,3',5,5'-tetramethylbenzidine (TMB) has been studied in the presence of 2-amino-4-nitrophenol (ANP), gallic acid (GA) or 4,4'-dihydroxydiphenylsulfone (DDS) and their polydisulfides poly(ADSNP), poly(DSGA), poly(DSDDS) at 20 degrees C in 10 mM
E I Karasyova et al.
Biochemistry. Biokhimiia, 66(6), 608-617 (2001-06-26)
The effects of different concentrations of 2-amino-4-nitrophenol (ANP) and of its polydisulfide (poly(ADSNP)) on peroxidase-catalyzed oxidation of 3,3',5,5'-tetramethylbenzidine (TMB) were studied at 20 degrees C in reversed micelles of AOT (0.2 M) in heptane and in mixed reversed micelles of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service