Skip to Content
Merck
All Photos(1)

Key Documents

A71328

Sigma-Aldrich

4-Aminophenol

≥98%

Synonym(s):

4-Hydroxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Beilstein:
385836
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

granular
powder or crystals

mp

185-189 °C (lit.)

solubility

water: slightly soluble

SMILES string

Nc1ccc(O)cc1

InChI

1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2

InChI key

PLIKAWJENQZMHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Aminophenol (4-Hydroxyaniline) is an organic building block. Its quantification in water samples upto the detection limit of 8×10-10mol l-1 has been proposed by employing single-wall carbon nanotubes (SWNT)-nafion film coated glassy carbon electrodes. It is present as the main contaminant in pharmaceutical formulations of paracetamol. High-performance liquid chromatographic (HPLC) method with amperometric detection has been reported for its determination in various analgesic formulations. It has been reported to be formed from the reduction of 4-nitrophenol (Nip) under metal-free conditions catalyzed by N-doped graphene (NG).

Application

4-Aminophenol is suitable for use in the synthesis of 2,2-bis(4-aminophenoxy) benzonitrile [4-APBN], a monomer required for the preparation of series of polyamides and poly(amide-imide)s. It may be used as derivatization reagent to improve the ionization of aliphatic and aromatic aldehydes by paper spray ionization mass spectrometry.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-free catalytic reduction of 4-nitrophenol to 4-aminophenol by N-doped graphene.
Kong XK, et al.
Energy & Environmental Science, 6(11), 3260-3266 (2013)
Saxena A, et al.
Eur. Polymer J., 39(2), 401-405 (2003)
E Wyszecka-Kaszuba et al.
Journal of pharmaceutical and biomedical analysis, 32(4-5), 1081-1086 (2003-08-06)
A method for the determination of 4-aminophenol, the main impurity of paracetamol, by high-performance liquid chromatographic (HPLC) method with amperometric detection has been developed. The analysis was performed in an isocratic mode on a reversed phase Luna column 5 microm
Wensheng Huang et al.
Talanta, 61(3), 411-416 (2008-10-31)
A single-wall carbon nanotubes (SWNT)-Nafion film coated glassy carbon electrode (GCE) was described for the determination of 4-aminophenol. In pH 3.0 sodium citrate-HCl buffer, the oxidation peak current of 4-aminophenol increases greatly at the SWNT-Nafion film coated GCE in contrast
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(14), 3906-3910 (2012-02-14)
We describe here the ability of manganese oxide monosheets to aggregate to form layered structures with 4-aminophenol molecules. These aggregated monosheets could be considered as the first step to synthesize a self-assembled layered hybrid of phenol-manganese ions with phenol and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service