Skip to Content
Merck
All Photos(1)

Key Documents

938203

Sigma-Aldrich

Cobalt iron oxide

nanoparticles, 8 nm particle size, 1 wt. % in water

Synonym(s):

CoFe2O4 nanoparticles, Cobalt ferrite nanoparticles, Cobalt diiron tetraoxide, Cobalt ferrate, Cobalt ferrite, Cobalt iron spinel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CoFe2O4
CAS Number:
Molecular Weight:
234.62
MDL number:
UNSPSC Code:
12352303

Quality Level

form

dispersion

concentration

1 wt. % in water

particle size

8 nm

bp

>1000 °C (Cobalt iron oxide)

mp

>400 °C (Cobalt iron oxide)

SMILES string

[Co+2].[Co+2].[Fe+3].[Fe+3].[O-2].[O-2].[O-2].[O-2].[O-2]

InChI

1S/2Co.2Fe.5O/q2*+2;2*+3;5*-2

InChI key

XNGOCNMGDFPQTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our cobalt iron oxide nanoparticles are magnetic CoFe2O4 nanoparticles with an average particle size of 8 nm dispersed in a 1wt% concentration in an aqueous solution. They boast a specific magnetization of >40 emu/g.

Application

Cobalt iron oxide nanoparticles are used in many applications because of their unique electrical, magnetic, and catalytic properties coupled with high biocompatibility. Magnetic nanoferrite particles been studied for important applications in biomedical research such as magnetic resonance imaging, targeted magnetic drug delivery, cell separation, and magnetic hyperthermia Cobalt iron oxide nanoparticles can also serve as catalysts in various chemical reactions due to their high surface area and unique electronic properties. For example, they have been investigated for applications in catalytic reactions, including water splitting, carbon dioxide reduction, and organic synthesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of cobalt ferrite nanoparticles in biomedical nanotechnology
Srinivasan Y S, et al.
Nanomedicine (London, England), 13(10), 1221-1238 (2018)
Low-Temperature Synthesis and Catalytic Activity of Cobalt Ferrite in Nitrous Oxide (N2O) Decomposition Reaction
Denisova K, et al.
Catalysts, 11(8), 889-889 (2021)
Magnetic Nanoparticles as MRI Contrast Agents
Avasthi A, et al.
Topics in Current Chemistry, 378(3), 40-40 (2020)
Cobalt nanoparticles as a novel magnetic resonance contrast agent--relaxivities at 1.5 and 3 Tesla
Parkes M L, et al.
Contrast media & molecular imaging, 3(4), 150-156 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service