Skip to Content
Merck
All Photos(3)

Documents

593044

Sigma-Aldrich

Aluminum nitride

nanopowder, <100 nm particle size

Synonym(s):

Aluminium nitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlN
CAS Number:
Molecular Weight:
40.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

particle size

<100 nm

mp

>2200 °C (lit.)

density

3.26 g/mL at 25 °C (lit.)

SMILES string

N#[Al]

InChI

1S/Al.N

InChI key

PIGFYZPCRLYGLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum nitride (AlN) is a wide bandgap semiconductor with exceptional thermal conductivity, high electrical insulation, and piezoelectric properties. It is widely used in electronics, such as in high-power and high-frequency devices, as well as in optoelectronics for UV light emitters. Additionally, AlN is utilized in biomedical devices due to its biocompatibility.

Application

  • This review explores the significant properties of aluminum nitride in the context of semiconductor materials, discussing its large bandgap and wide transparency range, which are advantageous for ultraviolet to mid-infrared applications (Li et al., 2021).

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphologically selective synthesis of nanocrystalline aluminum nitride.
Haber JA, et al.
Chemistry of Materials, 10(12), 4062-4071 (1998)
Shuangxi Nie et al.
Nanomaterials (Basel, Switzerland), 9(8) (2019-08-07)
Nanocomposites consisting of cellulose nanofibrils (CNFs) and nano-aluminum nitride (AlN) were prepared using a simple vacuum-assisted filtration process. Bleached sugarcane bagasse pulp was treated with potassium hydroxide and sodium chlorite, and was subsequently ultra-finely ground and homogenized to obtain CNFs.
Nanocrystalline aluminum nitride: II, sintering and properties.
Panchula ML and Ying JY
American Chemical Science Journal, 86(7), 1121-1127 (2003)
Electrical and thermophysical properties of epoxy/aluminum nitride nanocomposites: Effects of nanoparticle surface modification
Peng W, et al.
Composites Part A: Applied Science and Manufacturing, 41(9), 1201-1209 (2010)
Chengjie Zuo et al.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control, 57(1), 82-87 (2009-12-31)
This paper reports on the first demonstration of a 1.05-GHz microelectromechanical (MEMS) oscillator based on lateral-field-excited (LFE) piezoelectric AlN contourmode resonators. The oscillator shows a phase noise level of -81 dBc/Hz at 1-kHz offset frequency and a phase noise floor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service