Skip to Content
Merck
All Photos(2)

Key Documents

938149

Sigma-Aldrich

Ethyl methyl carbonate

greener alternative

Technipur®, 99.9%, ≤10 ppm acid, ≤10 ppm H2O

Synonym(s):

Carbonic acid ethyl methyl ester, EMC, Methyl ethyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
MDL number:
UNSPSC Code:
12191602
Assay:
99.9%
grade:
battery grade
form:
liquid

grade

battery grade

Quality Level

Assay

99.9%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤10 ppm H2O
≤10 ppm acid

bp

101 °C

mp

-55 °C

density

1.01 g/cm3

application(s)

battery manufacturing

greener alternative category

SMILES string

CCOC(=O)OC

InChI

1S/C4H8O3/c1-3-7-4(5)6-2/h3H2,1-2H3

InChI key

JBTWLSYIZRCDFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. EMC is an enabling product, it increases the solubility of lithium ions, improving the capacity density and power of batteries. Additionally, it has excellent thermal conductivity and low electrical resistance, which extends the working temperature range of electrolytes, enhances lithium battery safety, and prolongs their service life. EMC plays a crucial role in enhancing battery performance. Click here for more information.

Application

Ethyl methyl carbonate(EMC) is used as solvents for lithium ion battery electrolytes Linear alkyl carbonates as battery grade electrolytes having extremely low water (<10 ppm) and acid (<10 ppm) contents are critical for achieving high electrochemical performance

Legal Information

Product of MU Ionic Solutions Corp
Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Low-Temperature Electrolyte Using Isoxazole as the Main Solvent for Lithium-Ion Batteries
Tan S, et al.
ACS Applied Materials & Interfaces, 13, 24995-25001 (2021)
Reaction Product Analyses of the Most Active ?Inactive? Material in Lithium-Ion Batteries?The Electrolyte. I: Themal Stress and Marker Molecules
Henschel J, et al.
Chemistry of Materials, 31, 9970-9976 (2019)
Investigation of the effect of dimethyl methylphosphonate (DMMP) on flame extinction limit of lithium-ion battery electrolyte solvents
Chen Z, et al.
Fuel: The Science and Technology of Fuel and Energy, 270, 117423-117423 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service